missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Color Coded Mercury Free pH Buffer, pH 10.00, Reagecon™
Click to view available options
Quantity:
1000 mL
Description
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.
Specifications
Color | Blue |
pH | 10.00 |
Physical Form | Liquid |
Quantity | 1000 mL |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'