missing translation for 'onlineSavingsMsg'
Learn More

Type I Borosilicate Glass Standard Opening Sample Vial with Crimp Top

Use these vials for a variety of applications. JG Finneran™ Type I Borosilicate Glass Standard Opening Sample Vials are designed to work in Agilent and other autosamplers.

Brand:  JG Finneran Associates 320111232

Code : 17

Additional Details : Weight : 0.25380kg

Product Code. 10687171

Quantity Price
1 £21.35 / Pack of 100
10 £15.91 / Pack of 100
In Stock 26

Add to basket


Description

Description

  • Neck finish allows use of patented Poly Crimp™ Seals or standard aluminum seals
  • Choose from clear or amber Type I borosilicate glass
  • Available with graduated marking spots
Specifications

Specifications

Borosilicate Glass (Type I)
11mm
1000Pack
100Pack
Standard Opening Vial
12 mm
2 mL
32mm
100Pack
11mm
Clear
Videos
Documents