Learn More
Thermo Scientific™ Cel-Line Epoxy Diagnostic Slides
These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Made of extra-white soda-lime glass with very low iron content. Clean, flat and suitable for immediate use
£104.87 - £107.00
Specifications
Length (Metric) 75mm
Width (Metric) 25mm
Thickness (Metric) 1mm
View More Specs
Products 3
Product Code Brand Diameter (Metric) Color Well Count Price Quantity & Availability  
Product Code Brand Diameter (Metric) Color Well Count Price Quantity & Availability  
15314474 View Documents Thermo Scientific™
X8X30#406ANCE
15mm Brown 2 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
15393352 View Documents Thermo Scientific™
X8X30#341ABCE
20mm Black 1 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
12321263 View Documents Thermo Scientific™
X8X30#66A#BCE
6mm Black 8 This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Binds and keeps liquids applied to the well area
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
Specifications
Specifications
75mm
25mm
1mm