missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Set of Bent Hose Barbs with GL14 threads for condensers
Contains hose barbs, cap nuts, seals.
Brand: BUCHI 037287
Code : 61
Additional Details : Weight : 0.01800kg
Description
- For use with Rotavapor™
- Hose barbs, set 4pcs, bent GL14, silicone seal
Specifications
R-300 | |
Hose Barb | |
Silicone | |
Connection: GL14 |