Learn More
BRAND™ 384-Well PCR Microplate
Extra-thin and even wall thicknesses allow for optimal heat transfer and short cycle times. The reinforced top plate holds the PCR plate flat even during heating, making it easier to use in automated systemssuch as pipetting robots.
Brand: BRAND™ 781345
300.26 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Code : LB
Additional Details : Weight : 0.84000kg
Description
- Flexible, full-Skirted
- Free from DNase, DNA, RNase, and endotoxins.
- Extra-thin and even wall thicknesses allow for optimal heat transfer and short cycle times.
- Smooth shaft surfaces minimize enzyme and nucleic acid binding to the surface ensuring the reaction can run in full.
- The reinforced top plate holds the PCR plate flat even during heating, making it easier to use in automated systems such as pipetting robots.
- The edges of the wells are shaped so as to protect against cross-contamination and provide a reliable seal with sealing foil matched to the plates.
- The plates can be filled using multichannel pipettes or robotic systems.
- The ultra thin-walled PCR plate design facilitates constant, rapid and precise heat transfer.
- The smooth vessel interior minimizes the binding of enzymes and nucleic acid to the walls.
- The raised lips of the wells protect against cross-contamination, allowing a reliable seal with the sealing mat tailored to the plates.
Specifications
Standard | |
Autoclavable | |
384 | |
PCR | |
50/Pk. |
16 x 24 | |
Polypropylene | |
Yes | |
50 | |
Yes |