Learn More
Fisherbrand™ Safe-Seal™ Borosilicate Homogenizers
For use during the initial stages of sample preparation to break down and disrupt a range of materials such as animal or plant tissues, foodstuffs and soil. Fisherbrand™ Safe-Seal™ Borosilicate Homogenizers are tissue grinders manufactured from durable borosilicate glass.
POA
Specifications
Color Clear
Material Borosilicate glass, PTFE
View More Specs
Description
Description
  • With a PTFE pestle
  • With stainless steel handle
  • With borosilicate glass mortar which is stoppered in PTFE to protect against splash-back during the grinding process
Specifications
Specifications
Clear
Borosilicate glass, PTFE