Learn More
Fisherbrand™ Borosilicate Glass Rimless Culture Tubes
Use these tubes for tissue culture and clinical chemistry. Fisherbrand™ Borosilicate Glass Rimless Culture Tubes are made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass.
£57.80 - £242.00
Specifications
Description Culture Tube
Design Type Plain
Material Borosilicate Glass
Disposable Yes
For Use With For tissue culture and clinical chemistry
View More Specs
Products 4
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Price Quantity & Availability  
12347279 View Documents Fisherbrand™
10mm £57.80
Pack of 1000 View alternative products
Save Up to 
»
In Stock 41
Add to basket
11527403 View Documents Fisherbrand™
13mm £77.30
Pack of 1000 View alternative products
Save Up to 
»
In Stock 489
Add to basket
11597403 View Documents Fisherbrand™
20mm £242.00
Pack of 500 View alternative products
Save Up to 
»
In Stock 25
11537403 View Documents Fisherbrand™
15mm £70.95
Pack of 1000 View alternative products
Save Up to 
»
In Stock 9
Description
Description
  • Premium quality tubes with sturdy, uniform bottoms and consistent lengths.
  • Approximate wall thickness is 0.6mm
Specifications
Specifications
Culture Tube
Borosilicate Glass
For tissue culture and clinical chemistry
Plain
Yes
No