Learn More
Fisherbrand™ Borosilicate Glass Light-Walled Culture Tubes
Use these tubes for cell washing procedures. Fisherbrand™ Borosilicate Glass Light-Walled Culture Tubes are made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda lime glass.
£52.83 - £129.88
Specifications
Description Culture Tube
Design Type Uniform bottom
Material Borosilicate Glass
Disposable Yes
For Use With Suitable for cell washing procedures
View More Specs
Products 5
Product Code Brand Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Outer Length (Metric) Price Quantity & Availability  
11517403 View Documents Fisherbrand™
12mm 75mm £52.83
Pack of 1000 View alternative products
Save Up to 
»
In stock, ready to ship: 217
Add to basket
11547403 View Documents Fisherbrand™
16mm 100mm £78.65
Pack of 1000 View alternative products
Save Up to 
»
In stock, ready to ship: 406
Add to basket
11557403 View Documents Fisherbrand™
16mm 125mm £99.48
Pack of 1000 View alternative products
Save Up to 
»
In stock, ready to ship: 36
Add to basket
11577403 View Documents Fisherbrand™
16mm 150mm £111.17
Pack of 1000 View alternative products
Save Up to 
»
In stock, ready to ship: 96
Add to basket
11587403 View Documents Fisherbrand™
18mm 150mm £129.88
Pack of 500 View alternative products
Save Up to 
»
In stock, ready to ship: 129
Add to basket
Description
Description
  • Premium quality tubes with sturdy, uniform bottoms and consistent lengths.
  • Approximate wall thickness is 0.6mm
Specifications
Specifications
Culture Tube
Borosilicate Glass
Suitable for cell washing procedures
Uniform bottom
Yes