Learn More
Fisherbrand™ Borosilicate Mini Homogenizers
For use during the initial stages of sample preparation to break down and disrupt a range of materials such as animal or plant tissues, foodstuffs and soil. Fisherbrand™ Borosilicate Mini Homogenizers are tissue grinders manufactured from durable borosilicate glass.
£48.03 - £119.00
Specifications
Color Clear
Material Borosilicate glass
View More Specs
Products 3
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11522443 View Documents Fisherbrand™
1.0mL £48.03
Each View alternative products
Save Up to 
»
Estimated Shipment
22-04-2020
Add to basket
11512443 View Documents Fisherbrand™
0.1mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11532443 View Documents Fisherbrand™
3mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Ideal for micro tissue work
  • Bodies are clear precision glass
  • Pestles have ground glass working length
Specifications
Specifications
Clear
Borosilicate glass