Learn More
Fisherbrand™ Borosilicate Dounce Homogenizers
Designed for fine particle size reductions without damage to cell nuclei. Fisherbrand™ Borosilicate Dounce Homogenizers are tissue grinders for use during the initial stage of sample preparation.
£283.00 - £394.00
Specifications
Color Clear
Material Borosilicate glass
View More Specs
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
11572443 View Documents Fisherbrand™
7mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11582443 View Documents Fisherbrand™
15mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11562443 View Documents Fisherbrand™
1mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
11592443 View Documents Fisherbrand™
40mL This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
N/A View alternative products
Save Up to 
»
Description
Description
  • Supplied with two interchangeable pestles ground and polished for use in same tube
  • "Loose" pestle used for sample preparation, "Tight" pestle for final homogenate
  • Manufactured from durable borosilicate glass
Specifications
Specifications
Clear
Borosilicate glass