missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Bismuth(III) fluoride, 99.999% (metals basis)

Brand:  Alfa Aesar™ 011844.14

Code : NEW

Additional Details : CAS Number : 7787-61-3 Weight : 0.02500kg Transport : UN number : 3260 Chem class : 8 Pack group : III


Quantity 25g

 View more versions of this product

Product Code. 11391908

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance


Specifications

Chemical Identifiers

7787-61-3
265.976
BRCWHGIUHLWZBK-UHFFFAOYSA-K
82233
F[Bi](F)F
BiF3
MFCD00010893
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
trifluorobismuthane

Specifications

Bismuth(III) fluoride
(metals basis)
MFCD00010893
14,1265
BRCWHGIUHLWZBK-UHFFFAOYSA-K
trifluorobismuthane
82233
Powder
−200 Mesh
1.74
727°C
7787-61-3
BiF3
UN3260
bismuth fluoride, bismuth iii fluoride, bismuthine, trifluoro, trifluorobismuthine, bismuth trifluoride, bismuth iii trifluoride, acmc-1bd9n, ksc378g8f
F[Bi](F)F
265.976
265.98
99.999%
Odorless
Hygroscopic
25g
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Bismuth(III) fluoride

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS Documents