missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Agilent BioTek ELx50 Microplate Strip Washer
Syringe-drive fluid delivery system for precise control
POA
Specifications
Certifications/Compliance | CE and ETL marked |
---|---|
Automation Interface | RS-232 |
Depth (English) | 16 in. |
Height (English) | 6.5 in. |
Includes | QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit |
Includes: QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit
Description
Key Features
- Washes single strips or full plates
- Efficient washing ensures low background absorbance and small residue volumes
- Self-contained and programmable
- Micro-controlled, syringe-drive positive-displacement pump fluid delivery system for precise control
- Biomagnetic separation and vacuum filtration options available
- Safety features include aerosol cover, built-in decontamination routine, and isolation of electronics from fluidics
- Biomagnetic separation and vacuum filtration options available
- Choice of 8-, 12- or 16-channel manifold models
- Optional switching of up to three wash buffers
Applications:
Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays
Certifications
CE listed; ETL listed
Specifications
CE and ETL marked | |
16 in. | |
QC report and performance verification data sheet; Aerosol cover; Built-in pumps; Accessory package including bottles, tubing, and maintenance kit | |
14 in. | |
150 to 1000 μL/well/sec. | |
100/240 V |
RS-232 | |
6.5 in. | |
1 to 10 | |
LCD | |
Suitable for EIA, fluorescence, chemiluminescence, RIA, DNA probe, and cellular assays | |
20 lb. |
For in vitro diagnostic use