Learn More
Thermo Scientific™ Armadillo™ 384-Well PCR Plates
Optimize high throughput robotic PCR and qPCR applications with these uItra-rigid 384-well PCR plates with poylcarbonate frames and polypropylene wells.
£294.00 - £331.00
Specifications
Volume (Metric) Well 40 μL
Material Polypropylene
Volume (Metric) Working 25 μL
View More Specs
Products 12
Product Code Brand Color Well Color Price Quantity & Availability  
Product Code Brand Color Well Color Price Quantity & Availability  
12677516 View Documents Thermo Scientific™
BC2384
Clear Clear £294.00
Pack of 50 View alternative products
Save Up to 
»
EU Stock 1
Add to basket
12687516 View Documents Thermo Scientific™
BC2384B
Blue Clear £298.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12697516 View Documents Thermo Scientific™
BC2384G
Green Clear £298.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12607526 View Documents Thermo Scientific™
BC2384O
Orange Clear £298.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12677396 View Documents Thermo Scientific™
BC2384R
Red Clear £299.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12607446 View Documents Thermo Scientific™
BC2384Y
Yellow Clear £298.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12657526 View Documents Thermo Scientific™
BC3384
Clear White £331.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 02-03-2020
Add to basket
12661005 View Documents Thermo Scientific™
BC3384B
Blue White £330.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12667526 View Documents Thermo Scientific™
BC3384G
Green White £330.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12677526 View Documents Thermo Scientific™
BC3384O
Orange White £330.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12687396 View Documents Thermo Scientific™
BC3384R
Red White £329.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
12617466 View Documents Thermo Scientific™
BC3384Y
Yellow White £329.00
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 09-03-2020
Add to basket
Description
Description
Highlights
  • Optically clear skirt - Aids in visualization of liquids and improves stacking capabilities in automated handling systems
  • Raised rim design - Improved well surface area improves seal integrity in automated heat sealing
  • ANSI:SBS footprint - With rigidity of polycarbonate skirt, the ANSI footprint ensures complete compatibility with robotic systems for high-throughput applications
  • Thermostable polycarbonate frame design - Warp-resistant thermal cycling and enhanced mechanical stability in robotic handling
  • Thin walled U-bottom wells - Optimal heat transfer and maximum sample recovery
  • Indented Alphanumeric Grid reference lettering - Integrated frame and well design for improved heat sealing
  • Low profile design - Allows for maximum heat transfer and minimal evaporation when utilizing low-volume samples (30 µL working volume)
  • Available in both clear and white colored wells - Ensures the highest level of sensitivity for all PCR/qPCR reactions
  • Barcoding available - All plates are available already barcoded and can be provided barcoded to customer specifications for fully-automated applications
Specifications
Specifications
40 μL
Polypropylene
25 μL
This product is sold under license from Eppendorf AG.