Learn More
Path-ID™ Path-ID™ Multiplex One-Step Kit
The Path-ID™ Multiplex One-Step RT-PCR Kit is designed for multiplex, quantitative, reverse transcription PCR (qRT-PCR).
POA
Specifications
Detection Method | Primer-Probe |
---|---|
Form | Liquid |
PCR Method | 1-step RT-qPCR, Multiplex qPCR |
Reaction Speed | Standard |
For Use With (Equipment) | 7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT System, AB 7900 HT 384-Well System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode |
Includes: 1.375mL of 2X Multiplex RT-PCR Buffer, 280μL of 10X Multiplex Enzyme Mix including AmpliTaq Gold and ArrayScript, 1.75mL of Nuclease-free water
Description
It is optimized for the amplification of up to four targets simultaneously using your RNA samples and TaqMan™ primer/probe sets. Reactions are run using a single-tube, one-step procedure to reverse-transcribe the RNA and amplify your targets. The kit contains a Multiplex Enzyme Mix with ArrayScript™ reverse transcriptase and AmpliTaq Gold™ DNA Polymerase, and a buffer mix.
Multiplex One-Step Kit is optimized for amplification of up to four targets simultaneously using your RNA samples and TaqMan™ primer/probe sets. Reactions are run using a single-tube, one-step procedure to reverse-transcribe the RNA and amplify your targets. Kit contains a Multiplex Enzyme Mix with Arrayscript™ reverse transcriptase and AmpliTaq Gold™ DNA Polymerase, and a buffer mix.
Applications:
One-Step qRT-PCR, PCR & Real-Time PCR, Real Time PCR (qPCR)
Order Info
Built-In Hot Start. Guaranteed minimum shelf life is 60 days.
Specifications
Primer-Probe | |
1-step RT-qPCR, Multiplex qPCR | |
7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT System, AB 7900 HT 384-Well System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode | |
Path-ID™ | |
ArrayScript™ | |
Built-In Hot Start |
Liquid | |
Standard | |
ROX (Pre-mixed) | |
One-Step RT-PCR Kit | |
Pathogen Detection, Virus Detection | |
Total RNA, Viral RNA, mRNA |
For Research Use Only. Not for use in diagnostic procedures.