missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ AmpliTaq™ DNA Polymerase with Buffer I
A 94 kDa, thermostable, recombinant DNA polymerase
£163.00 - £9060.00
Specifications
Detection Method | Primer-probe |
---|---|
Format | Stand-alone enzyme |
Polymerase | AmpliTaq DNA Polymerase |
Reaction Speed | Standard |
Exonuclease Activity | 5' - 3' |
Includes: Contents: 5 x 200μL of AmpliTaq DNA Polymerase (5U/μL); 4 x 1.5mL of GeneAmp 10X PCR Buffer II (500mM KCl, 100mM Tris-HCl pH 8.3); 4 x 1.5mL of 25mM MgCl2.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10147874
|
Applied Biosystems™
N8080155 |
5000 Units |
£2070.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10468254
|
Applied Biosystems™
N8080171 |
1000 Units |
£542.00
Each |
Estimated Shipment: 08-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10516375
|
Applied Biosystems™
N8080185 |
25,000 Units |
£9060.00
Each |
Estimated Shipment: 15-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10458064
|
Applied Biosystems™
N8080152 |
3000 Units |
£1396.00
Each |
Estimated Shipment: 08-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10585025
|
Applied Biosystems™
N8080160 |
250 Units |
£163.00
Each |
Estimated Shipment: 05-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Recombinant DNA polymerase obtained by expression of modified form of Thermus aquaticus (Taq) DNA polymerase gene in E. coli. It is most thoroughly characterized enzyme available for PCR process and its recombinant nature and purification method provide unparalleled purity and reproducibility.
- Most thoroughly characterized enzyme available for PCR process, a testimony to its overall utility and efficacy
- Thermal activity profile makes it good for PCR applications
- QC-tested to guarantee reproducible results
PCR, PCR and Real-Time PCR, Routine PCR
Order Info
Shipping Condition: Dry ice
Specifications
Primer-probe | |
AmpliTaq DNA Polymerase | |
5' - 3' | |
DNA Polymerase | |
1 X | |
3'-A | |
5 kb or less |
Stand-alone enzyme | |
Standard | |
AmpliTaq™ | |
5 U/μL, 10X | |
No | |
Separate Components | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.