Learn More
IBA Solutions For Life Science™ ApG-dinucleotide IBA
Dinucleotides are used by RNA polymerases as initiators for RNA synthesis. Therefore, they are very useful for transcription analysis.
Brand:  IBA Solutions For Life Science™ 5-0710-116
Code : LE
Additional Details : Weight : 0.10000kg
Product Code. 12859356
  • £768.86 / 5mg
Estimated Shipment
24-06-2020