Learn More
PI16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534891
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (81%), Rat (83%), Pig (84%), Rhesus Monkey (96%), Chimpanzee (99%), Bovine (87%). IHC notes, Requires antigen retrieval using heat mediated 10mM Citrate buffer (pH6.0) or Tris-EDTA buffer (pH8.0) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
PI16 Polyclonal antibody specifically detects PI16 in Human samples. It is validated for Western Blot.Specifications
PI16 | |
Polyclonal | |
Unconjugated | |
PI16 | |
1200009H11Rik; CD364; Cripi; CRISP9; CRISP-9; Cysteine-rich protease inhibitor; cysteine-rich secretory protein 9; microseminoprotein, beta-binding protein; MSMBBP; peptidase inhibitor 16; Pi16; PI-16; protease inhibitor 16; PSEC0164; PSP94-binding protein; PSPBP; UNQ289/PRO328 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221476 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.86 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6UXB8 | |
PI16 | |
Recombinant fragment corresponding to a region within amino acids 1 and 192 of PI16 (Uniprot ID#Q6UXB8) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |