Learn More
NTF3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA118387
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
NTF3 Polyclonal antibody specifically detects NTF3 in Chicken, Human, Rat samples. It is validated for ELISA, Immunohistochemistry (Frozen), Western Blot.Specifications
NTF3 | |
Polyclonal | |
Unconjugated | |
Ntf3 | |
AI316846; AI835689; HDNF; Nerve growth factor 2; neurotrophic factor; neurotrophin 3; neurotrophin-3; neurotrophin-3 (HDNF/NT-3); NGF2; NGF-2; NT3; NT-3; NTF3; Ntf-3 | |
Rabbit | |
100 μL | |
Primary | |
Chicken, Human, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Frozen), Western Blot | |
Conc. Not Determined | |
whole serum with no preservative | |
P18280, P20783, P25433 | |
Ntf3 | |
Synthetic peptide corresponding to residues Y(139) A E H K S H R G E Y(149) of human, mouse and rat NT3. | |
RUO | |
428099, 4908, 81737 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |