Learn More
Myc Tag Monoclonal Antibody (Myc.A7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA121316BTIN
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA1-21316 detects N-terminal or C-terminal Myc tagged fusion proteins. MA1-21316 may be stored at 4° C short term. Add 0.05% sodium azide if desired. For long term storage, store at -20° C, avoiding freeze/thaw cycles.
Myc Tag Monoclonal antibody specifically detects Myc Tag in Tag samples. It is validated for ChIP Assay, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Myc Tag | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P01106 | |
MYC | |
Synthetic peptide: EQKLISEEDL (Myc). | |
100 μg | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |
ChIP Assay, ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Myc.A7 | |
Unconjugated | |
MYC | |
46-0603; AU016757; avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; class E basic helix-loop-helix protein 39; cmyc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; MRTL; MYC; Myc protein; Myc proto-oncogene protein; Myc Tag; Myc2; MYCC; myc-related translation/localization regulatory factor; myelocytomatosis oncogene; Niard; Nird; Proto-oncogene c-Myc; R95025; R950-25; transcription factor p64; v-myc avian myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene homolog | |
Mouse | |
Protein A | |
RUO | |
4609 | |
Antibody | |
IgG1 |