Learn More
IL-10 Rat anti-Human, Unlabeled, Clone: JES3-12G8, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 559076
Additional Details : Weight : 0.01000kg
Description
The JES3-12G8 antibody reacts with human interleukin-10 (IL-10) and viral IL-10. The immunogen used to generate the JES3-12G8 hybridoma was human IL-10. This is a neutralizing antibody. This antibody is routinely tested by immunocytochemical analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Immunocytochemistry
Specifications
IL-10 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human IL-10 | |
0.25 mg | |
Immunology | |
Human, Virus | |
IgG2a |
Immunocytochemistry | |
JES3-12G8 | |
Unconjugated | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |