missing translation for 'onlineSavingsMsg'
Learn More

HSP90 beta Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA515031

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 10850791

  • £288.00 / 100µg

Estimated Shipment
27-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Heat shock proteins (HSP) are expressed in response to various biological stresses, including heat. HSP90 is a 90 kDa protein that is induced under stress conditions, but is also one of the most abundant cellular proteins found under non-stress conditions. HSP90 proteins are highly conserved molecular chaperones that have key roles in signal transduction, protein folding, protein degradation, and morphologic evolution. HSP90 proteins normally associate with other cochaperones and play important roles in folding newly synthesized proteins or stabilizing and refolding denatured proteins after stress. HSP90 has been found to be associated with a number of other intracellular proteins, including steroid receptors, actin, tubulin, aryl hydrocarbon (Ah) receptor, and some kinases.
Specifications

Specifications

HSP90 beta
Polyclonal
PBS with 0.09% sodium azide
P08238
D6S182; HSP90-BETA; HSP90B; HSPC2; HSPCB; 90-kda heat shock protein beta HSP90 beta; HSP 84; HSP 90; HSP84; heat shock 84 kDa; heat shock 90kD protein 1; beta; heat shock 90kDa protein 1; beta; heat shock protein HSP 90-beta; heat shock protein beta
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3326
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
HSP90AB1
Liquid
HSP90AB1
KLH conjugated synthetic peptide between 438-465 amino acids from the central region of human HSP90AB1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Videos
Documents