Learn More
Fyn Monoclonal Antibody (2A10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515865
Code : LD
Additional Details : Weight : 0.10000kg
Description
MA5-15865 targets FYN in indirect ELISA, FACS, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15865 immunogen is purified recombinant fragment of human FYN expressed in E. Coli. . MA5-15865 detects FYN which has a predicted molecular weight of approximately 61kDa.
Fyn Monoclonal antibody specifically detects Fyn in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot.Specifications
Fyn | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P06241 | |
Fyn | |
Purified recombinant fragment of human FYN expressed in E. Coli. | |
RUO | |
2534 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot | |
2A10 | |
Unconjugated | |
Fyn | |
AI448320; AW552119; C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; Fyn; fyn {ECO:0000312; FYN oncogene related to SRC, FGR, YES; Fyn proto-oncogene; FYN proto-oncogene, Src family tyrosine kinase; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Protooncogene Syn; proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; RGD:2641}; RP1-66H14.1; SLK; Src Kinase p59; Src like kinase; src/yes-related novel; src-like kinase; SYN; tyrosine kinase p59fyn(T); tyrosine-protein kinase Fyn | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |