Learn More
FXR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529853
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa, HepG2, mouse liver. Predicted reactivity: Mouse (99%), Rat (89%), Zebrafish (82%), Xenopus laevis (91%), Chicken (86%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
FXR1 Polyclonal antibody specifically detects FXR1 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
FXR1 | |
Polyclonal | |
Unconjugated | |
Fxr1 | |
1110050J02Rik; 9530073J07Rik; AI851072; FMR1 autosomal homolog 1; fragile X mental retardation autosomal homolog variant p1K; fragile X mental retardation autosomal homolog variant p2K; fragile X mental retardation autosomal homolog variant p4K; fragile X mental retardation autosomal homolog variant p5FK; fragile X mental retardation gene 1, autosomal homolog; fragile X mental retardation gene, autosomal homolog; fragile X mental retardation syndrome-related protein 1; fragile X mental retardation, autosomal homolog 1; fragile X mental retardation-related protein 1; fragile-X-related protein 1; Fxr1; Fxr1h; Fxr1p; hFXR1p; hypothetical protein LOC536793; mFxr1p | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14359, 8087 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.35 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P51114, Q61584 | |
Fxr1 | |
Recombinant protein fragment corresponding to a region within amino acids 77 and 378 of Human FXR1 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |