missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSTL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA524035
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with non-human primate based on sequence homology.
FSTL1 Polyclonal antibody specifically detects FSTL1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Specifications
FSTL1 | |
Polyclonal | |
Unconjugated | |
Fstl1 | |
AI316791; AW107808; FLJ50214; FLJ52277; follistatin like 1; follistatin-like 1; follistatin-like protein 1; Follistatin-related protein 1; FRP; FSL1; Fstl; FSTL1; MIR198; OCC1; OCC-1; TGF-beta-inducible protein TSC-36; tsc36; TSC-36 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
11167 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q12841 | |
Fstl1 | |
KLH conjugated synthetic peptide between 280-308 amino acids from the C-terminal region of human FSTL1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |