Learn More
FZD7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA14137
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-4137 detects mouse frizzled. PA1-4137 has been successfully used in Western blot procedures. By Western blot this antibody detects a ∽40 kDa protein representing mouse fz in mouse pancreas lysate. The PA1-4137 immunogen is a synthetic peptide corresponding to residues M(1) R G P G T A A S H S P C(12) of mouse Frizzled. This sequence is 91% conserved in rat.
FZD7 Polyclonal antibody specifically detects FZD7 in Mouse samples. It is validated for Western Blot.Specifications
FZD7 | |
Polyclonal | |
Unconjugated | |
FZD7 | |
frizzled 7, seven transmembrane spanning receptor; frizzled class receptor 7; frizzled family receptor 7; frizzled homolog 7; frizzled homolog 7 (Drosophila); Frizzled, drosophila, homolog of, 7; frizzled-7; Fz7; Fz-7; FZD7; FzE3; hFz7; LOC100360552; mFz7; rCG22430-like; rizzled 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14369 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q61090 | |
FZD7 | |
Synthetic peptide corresponding to residues M(1) R G P G T A A S H S P C(12) of mouse Frizzled. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |