missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frizzled-2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533544
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Frizzled-2 Polyclonal antibody specifically detects Frizzled-2 in Bovine, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Frizzled-2 | |
Polyclonal | |
Unconjugated | |
Fzd2 | |
AL033370; AW456835; Dfrizzled2; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 2; frizzled 2, seven transmembrane spanning receptor; frizzled class receptor 2; frizzled family receptor 2; frizzled homolog 2; frizzled homolog 2 (Drosophila); frizzled-10; Frizzled-2; Fz10; fz-10; Fz2; Fz-2; Fzd10; Fzd2; fzE2; hFz2; Mfz10; Mfz10a; mFz2; rFz2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100050809, 100337669, 100350521, 100763109, 2535, 538507, 57265, 64512 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q08464, Q14332, Q9JIP6 | |
Fzd2 | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human Frizzled 2. | |
50 μg | |
Primary | |
Bovine, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit, Rat | |
Antibody | |
IgG |