missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXA2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518169
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse and rat based on sequence homology.
FOXA2 Polyclonal antibody specifically detects FOXA2 in Human, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
FOXA2 | |
Polyclonal | |
Unconjugated | |
FOXA2 | |
FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN | |
Goat | |
Antigen affinity chromatography | |
RUO | |
25099, 3170 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P32182, Q9Y261 | |
FOXA2 | |
Synthetic peptide sequence (GVYSRPIMNSS) corresponding to the C-terminus amino acids of FOXA2 | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |