Learn More
Follicle Stimulating Hormone Monoclonal Antibody (P4G2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514695
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-14695 targets Follicle Stimulating Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14695 immunogen is human Follicle Stimulating Hormone. MA5-14695 detects Follicle Stimulating Hormone which has a predicted molecular weight of approximately 13 kDa. Product MA514695 is a smaller package size of MIF2709 (formerly sold as a Seradyn product).
Follicle Stimulating Hormone Monoclonal antibody specifically detects Follicle Stimulating Hormone in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot.Specifications
Follicle Stimulating Hormone | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P01225, Q60687 | |
FSHB | |
Human Follicle Stimulating Hormone | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
P4G2 | |
Unconjugated | |
FSHB | |
class 1 follicle-stimulating hormone beta polypeptide; class 2 follicle-stimulating hormone beta polypeptide; follicle - stimulating hormone subunit beta; follicle stimulating hormone beta; follicle stimulating hormone beta subunit; follicle stimulating hormone beta-subunit precursor; follicle stimulating hormone subunit beta; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; follicle-stimulating hormone beta-subunit; follicle-stimulating hormone subunit beta; follicule stimulating hormone, beta polypeptide; follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH; FSH-alpha; FSHB; FSH-B; Fshbeta; FSH-beta; HH24; LSH-alpha; prefollicle stimulating hormone beta chain; TSH-alpha | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
14308, 2488 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |