missing translation for 'onlineSavingsMsg'
Learn More

EphA5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA514581

Code : LE

Additional Details : Weight : 0.01000kg

Product Code. 10754111

  • £288.00 / 100µg

Estimated Shipment
27-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
Specifications

Specifications

EphA5
Polyclonal
PBS with 0.09% sodium azide
P54756, Q60629
Ephrin type-A receptor 5; Tyrosine-protein kinase receptor EHK-1; Eph homology kinase-1; Receptor protein-tyrosine kinase HEK7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
13839, 2044
Immunohistochemistry, Western Blot
Unconjugated
EPHA5
Liquid
EPHA5
KLH conjugated synthetic peptide between 942-972 amino acids from the C-terminal region of human EphA5
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Videos
Documents