missing translation for 'onlineSavingsMsg'
Learn More

DCP2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA526190

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 13240329

  • £288.00 / 100µg

Estimated Shipment
17-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

DCP2 is a key component of an mRNA-decapping complex required for removal of the 5-prime cap from mRNA prior to its degradation from the 5-prime end (Fenger-Gron et al., 2005).
Specifications

Specifications

DCP2
Polyclonal
PBS with 0.09% sodium azide
Q8IU60
NUDT20, hDpc, mRNA-decapping enzyme 2, nudix (nucleoside diphosphate linked moiety X)-type motif 20
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
167227
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
DCP2
Liquid
DCP2
KLH conjugated synthetic peptide between 144-173 amino acids from the central region of human DCP2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Videos
Documents