missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Alfa Aesar Al-23 Tube, Both Ends Open, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Brand:  Thermo Scientific Alfa Aesar 031566.EM

Al-23 Tube, Both Ends Open, O.D.: 15mm, I.D.: 10mm, Alfa Aesar™

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 11323977

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.