missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Adcell™ PTFE Diagnostic Slides

These hydrophobic coatings bind liquids applied to the wells and keeps fluids in the well area. Clean, flat and suitable for immediate use

Brand:  Epredia™ X5XER202WAD1

Additional Details : Weight : 0.50000kg

Product Code. 10344031

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Binds and keeps liquids applied to the well area
  • PTFE Construction
  • Provides wettable wells for staining and growing cell cultures
  • Prevents cross-contamination
  • Hydrophobic coatings are very durable and withstand the strongest stains
  • Autoclavable
Specifications

Specifications

ADCELL
12
25 mm
White
100/Pk.
PTFE
1 mm
75 mm
5 mm Well
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers