Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Cimarec+ and SuperNuova+ Series Hotplates
Add functionality to your Thermo Scientific™ Cimarec+ and SuperNuova+ Series Hotplates with these accessories.
£43.15 - £68.25
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15463660 View Documents Thermo Scientific™
CIC0001447
PT100 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers £43.15
Each View alternative products
Save Up to 
»
Estimated Shipment
08-01-2020
15493660 View Documents Thermo Scientific™
CIC0001449
Hastelloy PT1000 External probe, 0-400°C, Hastelloy, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers £68.25
Each View alternative products
Save Up to 
»
Call for latest availability
+44 (0)1509 555 500
15483660 View Documents Thermo Scientific™
CIC0001448
PT1000 External probe, 0-400°C, Stainless steel, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers £46.20
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-12-2019