missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Accessories for the LP Vortex Mixer

Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.

Brand:  Thermo Scientific™ 88880124

Code : NEW

Additional Details : Weight : 0.11000kg

 View more versions of this product

Product Code. 15208884

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Optimize flexibility and performance by using Thermo Scientific™ Accessories for LP Vortex Mixers.

  •  Warranty: 2 years
Specifications

Specifications

Tube Holder
25 mm
Tube Holder with Insert Retainer (dia. 25mm)
Vortex Mixer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.