Learn More
3M™ Disposable Protective Overshoe Cover 403
The 3M™ Disposable Overshoe covers 402 are disposable shoe covers designed to provide basic barrier protection to help protect footwear against light liquid splashes and hazardous dusts.
Brand: 3M™ 7000089721
Code : 91
Additional Details : Weight : 1.00000kg
Description
3M™ Overboot Covers are a reliable, disposable boot cover designed to help protect footwear against light liquid splashes and hazardous dusts. Offered as an accessory, the overboots are made from a microporous laminate material with an anti-static treatment.- Covers are designed with ties and elastic at the calf to help ensure a good fit
- Distinctive unisex design
- Integrated sideshields with adjustable lens angle and temple lengths
- Vertically vented browgard minimizes fogging