missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopropionic acid, 95%, Thermo Scientific Chemicals

Catalog Number p-155402
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
79-42-5
C3H6O2S
106.14
MFCD00004862
PMNLUUOXGOOLSP-UHFFFAOYSA-N
2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
62326
CHEBI:47872
2-sulfanylpropanoic acid
CC(C(=O)O)S
This item is not returnable. View return policy
79-42-5
C3H6O2S
106.14
MFCD00004862
PMNLUUOXGOOLSP-UHFFFAOYSA-N
2-mercaptopropionic acid, thiolactic acid, 2-mercaptopropanoic acid, 2-thiolactic acid, 2-thiolpropionic acid, alpha-mercaptopropionic acid, alpha-mercaptopropanoic acid, propionic acid, 2-mercapto, propanoic acid, 2-mercapto, fema no. 3180
62326
CHEBI:47872
2-sulfanylpropanoic acid
CC(C(=O)O)S

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 10°C to 14°C
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 102°C (16.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Iodimetry)
Packaging Glass bottle
Linear Formula CH3CH(SH)CO2H
Refractive Index 1.4804 to 1.4824
Quantity 100 g
Beilstein 03, 289
Fieser 15,307
Merck Index 15, 9493
Specific Gravity 1.19
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with alcohol,ether and acetone,miscible with benzol,petroleumether,chloroform,,chloroethane,diethylether
Formula Weight 106.14
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Mercaptopropionic acid, 95%
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'