missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 6000 F Plus

Catalog Number 15715703
263.15 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.


Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

DuPont™ Tychem™ 6000 F Plus garments offer excellent chemical permeation resistance to numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals, particulates, biohazards and certain chemical warfare agents, in an innovative design.

  • DuPont™ Tychem™ 6000 F Plus. Hooded coverall available in grey. Elasticated face, wrists, waist and ankles for an optimal fit. Double cuffs, knitted inner cuffs, thumb loops to prevent sleeves from riding up. Front entry with double zipper, double flap and adhesive tape at outer flap.
    Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents. 
    Tychem™ 6000 F garment are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
  • Certified according to tradeulation (EU) 2016/425
  • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
  • EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Front closure with double zipper, double flap, outer flap with adhesive tape
  • Double cuffs, thumb loops, knitted inner cuffs
TRUSTED_SUSTAINABILITY
Size 2X-Large
Gender Unisex
Material Tychem
Color Gray
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Thumb Loop, Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Double Zipper with Self-adhesive Double Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
Resistance Chemicals, Liquid, Particle, Anti-static, Abrasion
Collar Type Elastic
Certifications/Compliance Certified according to Regulation (EU) 2016/425, Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B, EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination), Antistatic treatment (EN 1149-5) - on inside
Description Tychem™ 6000 F
No. of Pockets None
Size (English) Chest 46-49 in.
Size (Metric) Chest 116 to 124 cm
Thickness 210 μm
For Use With (Application) chemical spill clean-up, emergency response, to military and petrochemical applications.
Height (Metric) 186 to 194 cm
Weight (English) 120 g/m2
Quantity 10/Cs.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'