missing translation for 'onlineSavingsMsg'
Learn More

Scilabware Tray mercury stoppered 400mm x 310mm x 19mm

Brand:  Scilabware TWR056

Tray mercury stoppered 400mm x 310mm x 19mm

Code : 02

Additional Details : Weight : 0.01000kg

Product Code. 10389494

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.